CO1H324A608A CombiPhos Catalysts Inc

(3-cyano-5-cyclopropylphenyl)boronic acid

  • Cas Number : 2225178-19-6
  • Molecular Weight : 187.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart