CO1H324A6096 CombiPhos Catalysts Inc

(2-cyclopropyl-4-fluorophenyl)boronic acid

  • Cas Number : 1824647-97-3
  • Molecular Weight : 179.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart