CO1H324A609B CombiPhos Catalysts Inc

(2-cyclopropyl-5-methylphenyl)boronic acid

  • Cas Number : 2225179-07-5
  • Molecular Weight : 176.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart