CO1H324A609C CombiPhos Catalysts Inc

(5-chloro-2-cyclopropylphenyl)boronic acid

  • Cas Number : 2225175-49-3
  • Molecular Weight : 196.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart