CO1H324A60A8 CombiPhos Catalysts Inc

(4-(piperidin-4-yl)phenyl)boronic acid

  • Cas Number : 1967833-52-8
  • Molecular Weight : 205.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart