CO1H324A60AB CombiPhos Catalysts Inc

(3-CYANO-5-METHOXYPHENYL)BORONIC ACID

  • Cas Number : 960589-15-5
  • Molecular Weight : 176.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart