CO1H324A60AF CombiPhos Catalysts Inc

3-Trifluoromethyl-2-cyclopropylphenylboronic acid

  • Cas Number : 2225175-32-4
  • Molecular Weight : 229.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart