CO1H324A60B4 CombiPhos Catalysts Inc

3-Amino-2-cyclopropylphenylboronic acid

  • Cas Number : 2225180-87-8
  • Molecular Weight : 177.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart