CO1H324A60B6 CombiPhos Catalysts Inc

2-Methoxy-6-cyclopropylphenylboronic acid

  • Cas Number : 2225169-89-9
  • Molecular Weight : 192.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart