CO1H324A60B9 CombiPhos Catalysts Inc

2-Fluoro-6-cyclopropylphenylboronic acid

  • Cas Number : 1373394-32-1
  • Molecular Weight : 179.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart