CO1H324A60C3 CombiPhos Catalysts Inc

3-Amino-5-bromophenylboronic acid

  • Cas Number : 2225179-12-2
  • Molecular Weight : 215.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart