CO1H324A60C5 CombiPhos Catalysts Inc

3-Fluoro-5-(imidazol-1-yl)phenylboronic acid

  • Cas Number : 2225155-88-2
  • Molecular Weight : 205.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart