CO1H324A60D5 CombiPhos Catalysts Inc

2-Bromo-5-(methoxymethyl)phenylboronic acid

  • Cas Number : 2225178-27-6
  • Molecular Weight : 244.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart