CO1H324A60DA CombiPhos Catalysts Inc

3-Methyl-5-(methoxymethyl)phenylboronic acid

  • Cas Number : 2225155-68-8
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart