CO1H324A60DE CombiPhos Catalysts Inc

(4-cyclopropyl-2-(methoxycarbonyl)phenyl)boronic acid

  • Cas Number : 1395826-22-8
  • Molecular Weight : 220.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart