CO1H324A60E0 CombiPhos Catalysts Inc

(3-cyclopropyl-4-(methoxycarbonyl)phenyl)boronic acid

  • Cas Number : 1422975-77-6
  • Molecular Weight : 220.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart