CO1H324A60ED CombiPhos Catalysts Inc

(3-fluoro-4-(1H-pyrazol-1-yl)phenyl)boronic acid

  • Cas Number : 1778667-03-0
  • Molecular Weight : 205.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart