CO1H324A60F0 CombiPhos Catalysts Inc

(4-(1H-imidazol-1-yl)-3-(trifluoromethyl)phenyl)boronic acid

  • Cas Number : 2256708-74-2
  • Molecular Weight : 255.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart