CO1H324A60F4 CombiPhos Catalysts Inc

(4-(3-methyl-1H-pyrazol-1-yl)-3-(trifluoromethyl)phenyl)boronic acid

  • Cas Number : 2256708-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart