CO1H324A60FA CombiPhos Catalysts Inc

(2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)boronic acid

  • Cas Number : 2225170-62-5
  • Molecular Weight : 250.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart