CO1H324A6101 CombiPhos Catalysts Inc

2-Bromo-4-(piperazin-1-yl)phenylboronic acid

  • Cas Number : 2225178-26-5
  • Molecular Weight : 284.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart