CO1H324A6102 CombiPhos Catalysts Inc

(4-(piperazin-1-yl)-2-(trifluoromethyl)phenyl)boronic acid

  • Cas Number : 2225169-50-4
  • Molecular Weight : 274.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart