CO1H324A6109 CombiPhos Catalysts Inc

(4-morpholino-2-(trifluoromethyl)phenyl)boronic acid

  • Cas Number : 2225174-23-0
  • Molecular Weight : 275.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart