CO1H324A610B CombiPhos Catalysts Inc

2-Cyano-4-(morpholino)phenylboronic acid

  • Cas Number : 2225179-08-6
  • Molecular Weight : 232.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart