CO1H324A6117 CombiPhos Catalysts Inc

2-Trifluoromethyl-4-(3-methylpiperidin-1-yl)phenylboronic acid

  • Cas Number : 2225153-43-3
  • Molecular Weight : 287.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart