CO1H324A6119 CombiPhos Catalysts Inc

2-Cyano-4-(3-methylpiperidin-1-yl)phenylboronic acid

  • Cas Number : 2225153-10-4
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart