CO1H324A6124 CombiPhos Catalysts Inc

(2-methoxy-4-(piperidin-1-yl)phenyl)boronic acid

  • Cas Number : 2225179-31-5
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart