CO1H324A6128 CombiPhos Catalysts Inc

2-Cyano-4-(piperidin-1-yl)phenylboronic acid

  • Cas Number : 2225178-31-2
  • Molecular Weight : 230.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart