CO1H324A6129 CombiPhos Catalysts Inc

2-Fluoro-4-(piperidin-1-yl)phenylboronic acid

  • Cas Number : 2225180-92-5
  • Molecular Weight : 223.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart