CO1H324A6130 CombiPhos Catalysts Inc

2-Fluoro-4-(pyrrolidino)phenylboronic acid

  • Cas Number : 1876473-45-8
  • Molecular Weight : 209.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart