CO1H324A6134 CombiPhos Catalysts Inc

2-Trifluoromethyl-4-(4-methylimidazol-1-yl)phenylboronic acid

  • Cas Number : 2225178-35-6
  • Molecular Weight : 270.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart