CO1H324A6135 CombiPhos Catalysts Inc

2-Chloro-4-(4-methylimidazol-1-yl)phenylboronic acid

  • Cas Number : 2225174-31-0
  • Molecular Weight : 236.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart