CO1H324A6145 CombiPhos Catalysts Inc

2-Fluoro-4-(imidazol-1-yl)phenylboronic acid

  • Cas Number : 2225172-36-9
  • Molecular Weight : 205.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart