CO1H324A6146 CombiPhos Catalysts Inc

2-Methyl-4-(imidazol-1-yl)phenylboronic acid

  • Cas Number : 2225169-72-0
  • Molecular Weight : 202.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart