CO1H324A6148 CombiPhos Catalysts Inc

2-Bromo-4-(1H-pyrrol-1-yl)phenylboronic acid

  • Cas Number : 2225169-01-5
  • Molecular Weight : 265.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart