CO1H324A614A CombiPhos Catalysts Inc

2-Chloro-4-(1H-pyrrol-1-yl)phenylboronic acid

  • Cas Number : 2225171-67-3
  • Molecular Weight : 221.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart