CO1H324A614B CombiPhos Catalysts Inc

2-Cyano-4-(1H-pyrrol-1-yl)phenylboronic acid

  • Cas Number : 2225176-08-7
  • Molecular Weight : 212.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart