CO1H324A6151 CombiPhos Catalysts Inc

2-Chloro-4-(1H-pyrazol-1-yl)phenylboronic acid

  • Cas Number : 2225180-95-8
  • Molecular Weight : 222.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart