CO1H324A6159 CombiPhos Catalysts Inc

2-Cyano-4-(3-methyl-1H-pyrazol-1-yl)phenylboronic acid

  • Cas Number : 2225176-16-7
  • Molecular Weight : 227.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart