CO1H324A6161 CombiPhos Catalysts Inc

2-Methyl-4-(4-methyl-1H-pyrazol-1-yl)phenylboronic acid

  • Cas Number : 2225172-61-0
  • Molecular Weight : 216.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart