CO1H324A616D CombiPhos Catalysts Inc

2,3-Dimethyl-5-cyclopropylphenylboronic acid

  • Cas Number : 2225178-53-8
  • Molecular Weight : 190.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart