CO1H324A616F CombiPhos Catalysts Inc

2-Methyl-3-methoxy-5-cyclopropylphenylboronic acid

  • Cas Number : 2225170-21-6
  • Molecular Weight : 206.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart