CO1H324A6175 CombiPhos Catalysts Inc

2-Fluoro-5-methyl-3-cyclopropylphenylboronic acid

  • Cas Number : 2225169-88-8
  • Molecular Weight : 194.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart