CO1H324A6178 CombiPhos Catalysts Inc

2-Chloro-3-methyl-5-cyclopropylphenylboronic acid

  • Cas Number : 2225178-56-1
  • Molecular Weight : 210.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart