CO1H324A617D CombiPhos Catalysts Inc

2-Chloro-5-trifluoromethyl-4-methylphenylboronic acid

  • Cas Number : 2225169-10-6
  • Molecular Weight : 238.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart