CO1H324A618A CombiPhos Catalysts Inc

2-Bromo-3-methyl-5-trifluoromethylphenylboronic acid

  • Cas Number : 2225169-16-2
  • Molecular Weight : 282.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart