CO1H324A618D CombiPhos Catalysts Inc

2,5-Dimethyl-3-cyclopropylphenylboronic acid

  • Cas Number : 2225176-23-6
  • Molecular Weight : 190.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart