CO1H324A618E CombiPhos Catalysts Inc

3-Bromo-4-cyclopropyl-6-methylphenylboronic acid

  • Cas Number : 2225169-24-2
  • Molecular Weight : 254.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart