CO1H324A6191 CombiPhos Catalysts Inc

3-Chloro-2-methyl-5-methoxyphenylboronic acid

  • Cas Number : 2225178-57-2
  • Molecular Weight : 200.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart