CO1H324A6196 CombiPhos Catalysts Inc

2-Chloro-4-bromo-5-cyclopropylphenylboronic acid

  • Cas Number : 2225172-94-9
  • Molecular Weight : 275.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart